Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell ActivArmr™ Winter Monkey Grip™ 23-173 Heavy Duty PVC-Coated Insulated Gloves

Catalog No. p-143783 Shop All Ansell Products
Change view
Click to view available options
Size:
10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-019299 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-019299 Supplier Ansell Supplier No. 23173100
This item is not returnable. View return policy

Textured PVC coating protects hands from abrasions and provides water, oil, and grease repellency. Foam insulation remains flexible, and helps to keep hands warm in cold applications.

  • Textured PVC vinyl coating protects hands from abrasions and provides water, oil, and grease repellency
  • Coating remains soft and flexible in cold conditions
  • Foam insulation helps to keep hands warm
  • Enhanced grip capabilities when handling heavy-duty objects
  • Curved fingers and wing thumb increase user comfort
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category II
Construction Cut and Sewn
Coating Material PVC
Color Orange
Cut Resistance Rating ANSI A1
Exterior Finish Fully Textured
Hand Specific
Material PVC
Product Type Heavy-duty Gloves
Size 10
EN388 Standard 3241B
For Use With (Application) Cold Storage, Equipment Repair, Metal Fabrication, and Warehousing
Glove Cuff Style Safety
Grip Design Textured
Liner Material Cotton
Lining Lined
Series 23-173
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.