Learn More
PIP™ Anser™ Semi-Rimless Safety Glasses
Catalog No. 19148031 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Lens Coating:
Anti-fog, Scratch-resistant
Scratch-resistant
Lens Tint:
Clear
Gray
Includes:
Adjustable Neck Cord
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating Lens Tint Includes
19-148-031 Anti-fog, Scratch-resistant Gray Adjustable Neck Cord
19-148-000 Scratch-resistant Clear -
19-148-001 Scratch-resistant Gray -
19-148-002 Anti-fog, Scratch-resistant Clear -
19-148-003 Anti-fog, Scratch-resistant Gray -
19-148-030 Anti-fog, Scratch-resistant Clear Adjustable Neck Cord
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-148-031 Supplier PIP™ Supplier No. 250AN10521
Add to Cart
Add to Cart
Semi-Rimless Safety Glasses feature wraparound temples with rubberized tips gently grip the wearer's head.
  • Lightweight, semi-rimless design provides all day user comfort
  • Sleek, sporty style with wraparound lens protection
  • Bayonet temples with flexible rubber tips offer a comfortable, secure fit
  • Soft rubber nose pads help keep the glasses in place
  • Supplied with an adjustable neck cord
  • 9 Base curve lens
  • Meets ANSI Z87.1+
Specifications
Lens Coating Anti-fog, Scratch-resistant
Shape Bayonet Temples, Wraparound Lens
Height (Metric) 3.8 cm
Lens Tint Gray
Product Type Safety Glasses
For Use With (Application) Protection from projectiles and impact in general use
Frame Color Black
Includes Adjustable Neck Cord
Temple Style Wraparound Temples with Rubber Tips
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate, PVC
UV Protection 99.99% UVA and UVB Light Absorption
Frame Type Semi-rimless
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Warning - Prop 65 - Reproductive Harm