Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ ANSI Type R Class 2 Expandable Two-Tone Mesh Breakaway Vest

Catalog No. p-7220394 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
2X-Large to 5X-Large
Medium to X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-046-484 Medium to X-Large
19-046-485 2X-Large to 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-046-484 Supplier PIP™ Supplier No. 3020590LYM/XL
Only null left
Add to Cart
Add to Cart

Type R/Class 2 apparel is necessary for workers exposed to traffic traveling over 25 mph and those who work against complex backgrounds. Ideal for construction, municipalities, shipyards, or anywhere that hi-vis apparel is required.

  • Breathable polyester mesh with 5 point breakaway
  • Off shoulder breakaway system provides additional safety around extreme traffic hazards, or while moving machinery and equipment
  • Adjustable hook & loop sides for a universal fit
  • 2 in. two-tone tape for optimum visibility
  • Breakaway zipper front closure
  • 1 outer chest radio/cell pocket with ID pocket
  • 1 inner chest pocket
  • 1 lower inner pocket

Specifications

Product Type High-visibility Vest
Construction 5 Point Breakaway/Expandable
Closure Type Zipper
Certifications/Compliance ANSI/ISEA 107-2015 Class 2 Type R
Color Hi-vis Yellow
Size Medium to X-Large
Material Polyester Mesh
Length (English) 28 in.
Length (Metric) 71 cm
Size (English) Chest 46 in.
No. of Pockets 3
Size (Metric) Chest 117 cm
Tape Type Two-Tone
For Use With (Application) Construction, Surveying, Shipyards, and Municipalities
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.