Learn More
Advanced ImmunoChemical, Inc. Recombinant Canine Parvovirus VP2 Antigen, 1 mg, > 90% purity, Capsid protein VP2, Lyophilized, Use in immunoassays
Supplier:  Advanced ImmunoChemical, Inc. 2CNT CANT 53 1MG
Encompass_Preferred
This recombinant canine parvovirus (CPV) VP2 antigen is provided by Advanced ImmunoChemical Inc. and is designed for use in in vitro assay development and research, particularly for the detection of CPV or the determination of CPV antibody titers in canine blood. This highly purified protein is suitable as a calibrator or standard in immunoassays.
  • High purity greater than 90% as determined by SDS-PAGE under reducing conditions.
  • Validated immunoreactivity confirmed through reaction with monoclonal antibodies specific to canine parvovirus.
  • Ideal for use as a calibrator or standard in immunoassays aimed at CPV immunodetection.
  • Useful for quantifying CPV antibody levels in canine blood.
Catalog No. NC3433057