Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ANT1/ANT2/ANT3/ANT4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109394
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109394 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109394 Supplier Invitrogen™ Supplier No. PA5109394
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GMLPDPKNVH IFVSWMIAQS VTAVAGLVSY PFDTVRRRMM MQSGRKGADI MYTGTVDCWR KIAKDEGAKA FFKGAWSNVL RGMGGAFVLV LYDEIKKYV.

ANT1/2/3/4 (SLC25A4, SLC25A5, SLC25A6, SLC25A31) are members of the mitochondrial carrier subfamily of solute carrier protein genes. The product of this gene functions as a gated pore that translocates ADP from the mitochondrial matrix into the cytoplasm. The protein is implicated in the function of the permability transition pore complex (PTPC), which regulates the release of mitochondrial products that induce apoptosis. The human genome contains several non-transcribed pseudogenes of this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANT1/ANT2/ANT3/ANT4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.65 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Slc25a5
Gene Accession No. P05141, P12235, P48962, P51881, Q05962, Q09073, Q3V132, Q5U680, Q70HW3, Q9H0C2
Gene Alias 1700034J06Rik; 2F1; 4930433D19Rik; 4933433F13Rik; AAC1; AAC2; Aac4; Adenine nucleotid translocator 2 fibroblast isoform (ATP-ADP carrier protein); Adenine nucleotid translocator 2, fibroblast isoform (ATP-ADP carrier protein); adenine nucleotide translocase 4; adenine nucleotide translocase-1; adenine nucleotide translocator 1; adenine nucleotide translocator 1 (skeletal muscle); adenine nucleotide translocator 1, skeletal muscle; Adenine nucleotide translocator 2; adenine nucleotide translocator 2 (fibroblast); adenine nucleotide translocator 2 fibroblast isoform (ATP-ADP carrier protein); adenine nucleotide translocator 2, fibroblast isoform (ATP-ADP carrier protein); adenine nucleotide translocator 4; adenine nucleotide translocator), member 31; adenine nucleotide translocator), member 31-like; ADP,ATP carrier protein 1; ADP,ATP carrier protein 2; ADP,ATP carrier protein 4; ADP,ATP carrier protein, fibroblast isoform; ADP,ATP carrier protein, heart/skeletal muscle; ADP,ATP carrier protein, heart/skeletal muscle isoform T1; ADP/ATP translocase 1; ADP/ATP translocase 2; ADP/ATP translocase 2, N-terminally processed; ADP/ATP translocase 4; Anc1; ANT; ANT 1; ANT 2; ANT 4; ANT1; ANT2; ANT4; AU019225; AW557176; COXPD28; D6Bwg0781e; heart/skeletal muscle ATP/ADP translocator; mANC1; mitochondrial adenine nucleotide translocator; mitochondrial S-adenosylmethionine transporter; MTDPS12; PEO2; PEO3; PEOA2; S-adenosylmethionine mitochondrial carrier protein; SAMC; Sfec; SFEC35kDa; Slc25a26; Slc25a31; Slc25a4; SLC25A5; Slc25a6; solute carrier family 25 (mitochondrial adenine nucleotide translocator) member 4; solute carrier family 25 (mitochondrial carrier, adenine nucleotide translocator), member 31-like; solute carrier family 25 (mitochondrial carrier, adenine nucleotide translocator), member 4; solute carrier family 25 (mitochondrial carrier, adenine nucleotide translocator), member 5; solute carrier family 25 (mitochondrial carrier, phosphate carrier), member 26; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 31; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 4; solute carrier family 25 (mitochondrial carrier; adenine nucleotide translocator), member 5; solute carrier family 25 (mitochondrial carrier; phosphate carrier), member 26; solute carrier family 25 (S-adenosylmethionine carrier), member 26; solute carrier family 25 member 26; Solute carrier family 25 member 31; solute carrier family 25 member 4; Solute carrier family 25 member 5; solute carrier family 25, member 26; sperm flagellar energy carrier protein; SZFULLNAME2; SZGENESYMBOL2; T1; T2; T3
Gene Symbols Slc25a26, Slc25a31, Slc25a4, Slc25a5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human ANT1 (NP_001142.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115286, 11739, 11740, 25176, 291, 292, 362403, 67582, 689108, 73333, 83447, 85333
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.