Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Anthraquinone-2-carbonyl Chloride 98.0+%, TCI America™
SDP

Catalog No. A050325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A050325G 25 g
A05035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A050325G Supplier TCI America Supplier No. A050325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6470-87-7
Molecular Formula C14H10ClN3O3S
Molecular Weight (g/mol) 335.76
MDL Number MFCD00059498
InChI Key QBUULMJMEMIBIA-UHFFFAOYSA-N
PubChem CID 15025286
IUPAC Name 1-(4-chlorophenyl)-3-(2-nitrobenzoyl)thiourea
SMILES [O-][N+](=O)C1=CC=CC=C1C(=O)NC(=S)NC1=CC=C(Cl)C=C1

Specifications

Melting Point 147°C
Color Yellow
Boiling Point 210°C
Quantity 25 g
UN Number 3261
Formula Weight 270.67
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Anthraquinone-2-carbonyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.