Learn More
Apexbio Technology LLC HyperFluor™ 594 Rabbit Anti-Goat IgG (H+L) Antibody 5X100ul
Supplier: Apexbio Technology LLC K33080.5
HyperFluor 594 Rabbit Anti-Goat IgG HplusL Antibody is a fluorophore-conjugated secondary antibody targeting goat IgG heavy and light chains It is designed to specifically bind to goat-derived primary antibodies facilitating detection in immunoassays HyperFluor 594 Rabbit Anti-Goat IgG HplusL Antibody exerts its biological activity by recognizing and binding to goat IgG enabling fluorescent labeling with emission at 617 nm upon excitation at 590 nm Based on these properties HyperFluor 594 Rabbit Anti-Goat IgG HplusL Antibody is applicable in immunocytochemistry immunohistochemistry and flow cytometry
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.