Learn More
Medchemexpress LLC Anthraquinone-d8 | 10439-39-1 | 216.26 | C14D8O2 | 10 MG

Supplier: Medchemexpress LLC HYN0354S10MG
Anthraquinone-d8 is the deuterium-labeled analogue of anthraquinone used as an internal standard and tracer for quantitative analysis by NMR, GC-MS, and LC-MS, and as a precursor for dye synthesis. It is supplied as a pre-weighed solid (light yellow to yellow) suitable for analytical and research use.
- Deuterium-labeled (d8) isotopologue for tracer and internal standard use.
- Molecular formula: C14D8O2; molecular weight: 216.26.
- CAS number: 10439-39-1 for unambiguous identification.
- Appearance: light yellow to yellow solid suitable for handling in dry form.
- Storage: stable at room temperature for extended storage; in solvent, store at low temperatures as specified.
- Applications: quantitative NMR, GC-MS, LC-MS, and precursor for dye formation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.