Learn More
Invitrogen™ Anthrax Lethal Factor Monoclonal Antibody (5F502.2)

Description
Suggested positive control: recombinant protein.
Specifications
Specifications
| Antigen | Anthrax Lethal Factor |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 5F502.2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene Alias | Anthrax LF |
| Host Species | Mouse |
| Immunogen | Synthetic peptide corresponding to amino acids 779-792 of the Anthrax LF protein (approx. 90 kD). |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.