Learn More
Invitrogen™ Anthrax Protective Antigen Polyclonal Antibody

Description
Suggested positive control: PA recombinant protein.
Specifications
Specifications
| Antigen | Anthrax Protective Antigen |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene Alias | Anthrax PA |
| Host Species | Rabbit |
| Immunogen | Synthetic peptides corresponding to amino acids 186-192 of the Anthrax PA protein (approx. 83 kDa). |
| Quantity | 200 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.