Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Lipoxygenase Mouse, Unlabeled, Clone: 33, BD

Catalog No. p-3855561 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610694 50 μg
BDB610695 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610694 Supplier BD Biosciences Supplier No. 610694
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

5-Lipoxygenase (5-LO) is the initial enzyme that converts arachidonic acid to leukotrienes, which are important inflammatory and vasoconstrictive metabolites. It is activated in response to a number of stimuli, such as differentiation and allergen challenges. The 5-LO gene, abundantly expressed in placenta, lung, and leukocytes, encodes a protein of 674 amino acids with an apparent molecular weight of 78kDa. 5-LO is a Ca2+ and ATP-dependent enzyme that translocates from the cytosol to either a nuclear or plasma membrane compartment following activation. A proline-rich domain of 5-LO (amino acids 566-577) has been identified as a binding site for the PTyr-binding protein, Grb2. This Grb2 site links tyrosine kinases with activation and redistribution of 5-LO. Furthermore, tyrosine kinase inhibitors increase the activity of 5-LO and block the enzyme's subcellular redistribution.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen 5-Lipoxygenase
Applications Western Blot
Classification Monoclonal
Clone 33
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.