Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5'-Lipoxygenase Rabbit anti-Human, Mouse, Rat, Rabbit, Polyclonal, MilliporeSigma™

Catalog No. AB3796MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3796MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3796MI Supplier MilliporeSigma Supplier No. AB3796
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-5′-Lipoxygenase Antibody detects level of 5′-Lipoxygenase and has been published and validated for use in ELISA, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 5'-Lipoxygenase
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin in PBS with 50% Glycerol and 0.02% sodium azide
Gene Symbols ALOX5; 5-lipoxygenase; 5LPG; MGC163204; LOG5; 5-LO; EC 1.13.11.34
Host Species Rabbit
Immunogen Purified human 5'-Lipoxygenase
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000698.2
Target Species Human, Mouse, Rabbit, Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.