Learn More
AdipoGen anti-8-Hydroxy-2'-deoxyguanosine [8-OHdG], mAb (N45.1)
SDP
Supplier:  AdipoGen JAIMOG020P
Encompass_Preferred
anti-8-Hydroxy-2'-deoxyguanosine [8-OHdG], mAb (N45.1), AdipoGen Life Sciences. Monoclonal Antibody. Recognizes 8-OHdG. Does not cross-react with 19 analogues of 8-OHdG (guanosine (G), 7-methylG, 6-SHG, 8-bromoG, dA, dC, dT, dI, dU, dG, O6-methyldG, 8-OHdA, guanine (Gua), O6-methylGua, 8-OHGua, uric acid, urea, creatine, creatinine). Minimal cross-reactivity with 8-sulfhydrylG and 8-OHG (less than 1%). Lyophilized. Contains 10mM PBS, pH7.4 containing 1.0% BSA. 8-hydroxy-2'-deoxyguanosine (8-OHdG) is a modified base that occurs in DNA due to attack by hydroxyl radicals (incl. singlet oxygen and direct photodynamic action) that are formed as byproducts and intermediates of aerobic metabolism and during oxidative stress.
Catalog No. 50-101-5749
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.