Learn More
United States Biological Corporation ANTI A-SYNUCLEIN C-TERM
Supplier:  United States Biological Corporation S950001E100UL
Encompass_Preferred
Synnuclein family of proteins is abundantly expressed in neuronal cytosol and presynaptic terminals. In vertebrates they are encoded by three different genes. Synucleins have been specifically implemented in three major diseases: Alzheimers (AD), Parkinsons (PD) and breast cancer. In AD, a peptide derived from alpha-synuclein forms an intrinsic component of plaque amyloid. In PD, an alpha-synuclein accumulates in Lewy bodies. An allele of alpha-synuclein has been linked to many familial cases of PD. In breast cancer increased expression of gamma synuclein correlates with the disease progression. Synucleins appear to be involved in the membrane plasticity in developing song control system of songbirds. Applications: Suitable for use in Immunohistochemistry. Other applications not tested. Recommended Dilutions: Immunohistochemistry (frozen/ paraffin): 1:1000 Optimal dilutions to be determined by the researcher. Storage and Stability: Lyophilized powder may be stored at -20 �C.
Catalog No. NC9078096
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.