Learn More
ECM Bioscience α1-Catenin (a.a. 143-153) Rabbit Polyclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience CP3431
Encompass_Preferred
This antibody was affinity purified using α1-Catenin (a.a. 143-153) peptide (without carrier). The antibody detects a 102 kDa* protein corresponding to the molecular mass of α1-Catenin on SDS-PAGE immunoblots of rat PC12 and mouse SYF cells. The immunogen was α1-Catenin synthetic peptide (coupled to KLH) corresponding to amino acid residues 143 to 153 in human α1-Catenin. This peptide sequence is highly conserved in rat and mouse α1-Catenin, and has some homology to α2-Catenin or α3-Catenin. Accession #P35221. Applications = WB, E; Species Reactivity = Hu, Rt, Ms, Ck; Size = 100 μl; Buffer = PBS, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = alphaE-catenin, catenin alpha1, catenin
Catalog No. 50-206-8685
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.