Learn More
American Research Products Inc Anti-Adeno-associated virus (AAV), VP1/VP2/VP3, 250 uL
Supplier:  American Research Products Inc 61084
Encompass
Mouse Monoclonal Antibody to Adeno-associated virus (AAV), VP1/VP2/VP3. Useful for immunolocalization studies of capsid formation in combination with monoclonal antibody A20 (03-61055/65155) and rabbit polyclonal antibody to AAV capsid proteins (03-61084). Reacts with free VP1, VP2 and VP3 and at a very reduced degree with assembled capsids. Recognizes all AAV-serogroups 1-6 (4 reacts weaker).
Catalog No. NC1702312
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.