Learn More
American Research Products Inc ANTI-AAV9 MOUSE MONOCLONAL
Supplier:  American Research Products Inc 615162
Encompass
anti-AAV9 (intact particle) mouse monoclonal, ADK9, Biotin ConjugateFor characterization of different stages of infection and very useful for the analysis of the AAV assembly process. ADK9 specifically reacts with intact adeno-associated virus particles, empty and full capsids. Recognizes a conformational epitope of assembled capsids. The antibody cannot be used for immunoblotting. Limited Use Label License: Research Use OnlyProduct is exclusively licensed to PROGEN Biotechnik GmbH. The use of these products for the development, manufacturing and sale of secondary products/derivatives which are based on the purchased products and/or which include the purchased product require a royalty based sub-license agreement.
Catalog No. NC3238590
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.