Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl-Histone H3 (Lys27) Rabbit anti-Human, Vertebrates, Yeast (S. cerevisiae), Polyclonal, MilliporeSigma™

Catalog No. p-5168661 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-360-SMI 10 μL
07-360-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-360-SMI Supplier MilliporeSigma Supplier No. 07360S
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-acetyl-Histone H3 (Lys27) Antibody is a rabbit polyclonal antibody for detection of Histone H3 acetylated on lysine 27. Also known as Anti-H3K27ac this antibody is published in peer reviewed journals and is specificity verified by dot blot (DB) and validated in Chimmunoprecipitation, ChIP-seq, western blot, DB, Mplex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen acetyl-Histone H3 (Lys27)
Applications Multiplex, Dot Blot, Immunoassay, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Immunoaffinity Purified immunoglobulin in 0.2M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide, and 5mg/ml BSA before the addition of glycerol to 30%
Gene Symbols HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886
Host Species Rabbit
Immunogen synthetic peptide corresponding to the sequence ARACKSA, in which ACK corresponds to acetyl lysine 27 of yeast histone H3
Purification Method Unpurified
Quantity 10 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_003493
Target Species Human, S. cerevisiae, Vertebrates, Yeast
Content And Storage −20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.