Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl-Histone H3 (Lys9) Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. p-5168667 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 μg
200 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-942-SMI 20 μg
06-942-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-942-SMI Supplier MilliporeSigma Supplier No. 06942S
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-acetyl-Histone H3 (Lys9) Antibody is a Rabbit Polyclonal Antibody for detection of Histone H3 acetylated on lysine 9. This purified Ab, also known as Anti-H3K9Ac, is specificity verified by dot blot (DB), published in peer reviewed journals, and validated in western blot, Chimmunoprecipitation, ChIP-seq, flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen acetyl-Histone H3 (Lys9)
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Dot Blot, Flow Cytometry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit IgG in buffer containing 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%.
Gene Symbols HIST1H3A; H3FA; HIST1H3B; H3FL; HIST1H3C; H3FC; HIST1H3D; H3FB; HIST1H3E; H3FD; HIST1H3F; H3FI; HIST1H3G; H3FH; HIST1H3H; H3FK; HIST1H3I; H3FF; HIST1H3J; H3FJ; HIST3H3; H3FT; H3T; MGC126886
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide (ARTKQTA RAcKSTG-C) corresponding to amino acids 1-12 of Histone H3.
Purification Method Protein A purfied
Quantity 20 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_003493
Target Species Eukaryote, Human, Mouse, Rat
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.