Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Actin Ab-5 Mouse, Unlabeled, Clone: C4/ACTIN, BD

Catalog No. p-3879246 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612656 50 μg
BDB612657 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB612656 Supplier BD Biosciences Supplier No. 612656
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Changes in cellular morphology, adhesion, and motility occur through the reorganization of the actin cytoskeleton. This reorganization of actin filaments results from interactions between actin and actin-binding proteins. Actin is a 42kDa protein that is known as G-actin in its monomeric form. Polymerization of G-actin monomers leads to the generation of flexible filaments, 5-9 nm in diameter, called F-actin. F-actin may be organized in linear bundles called stress fibers or in two-dimensional networks. The latter are highly concentrated beneath the plasma membrane and form the actin cortex. Regulation of actin cytoskeletal dynamics occurs through actin-binding proteins. These proteins bind to G- and/or F-actin and regulate various aspects of actin cytoskeletal dynamics, such as polymerization and depolymerization of actin, cross-linking of actin filaments into bundles, interaction of actin-based structures with membranes and other cytoskeletal elements, and locomotion of actin-based structures. Thus, the actin cytoskeleton is a complex matrix consisting of G- and F-actin along with the multitude of interactions between these actin forms and a variety of different types of actin-binding proteins. The C4 monoclonal antibody reacts with all known isoforms of actin in vertebrate muscle and non-muscle cells.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Actin Ab-5
Applications Western Blot
Classification Monoclonal
Clone C4/ACTIN
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Chicken gizzard muscle Actin
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.