Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Actin Mouse anti-Human, Chicken, Clone: JLA20, MilliporeSigma™

Catalog No. MABT219MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT219MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT219MI Supplier MilliporeSigma Supplier No. MABT219
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Actin using this Anti-Actin Antibody, clone JLA20 validated for use in Western Blotting, Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Actin
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone JLA20
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_001026234
Gene Symbols ACTA1, ACTA
Host Species Mouse
Immunogen Chicken cytoskeletal proteins
Purification Method Purified by ion exchange chromatography
Quantity 100 μL
Primary or Secondary Primary
Target Species Chicken, Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.