Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ADD1 Matched Antibody Pair, Abnova

Catalog No. 89017202 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-202 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-202 Supplier Abnova Corporation Supplier No. H00000118AP11
Add to Cart
Add to Cart

Includes

Rabbit MaxPab™ affinity purified polyclonal anti-ADD1 (100μg); Mouse monoclonal anti-ADD1, IgG1κ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.

Adducins are a family of cytoskeleton proteins encoded by three genes (α, β, γ). Adducin is a heterodimeric protein that consists of related subunits, which are produced from distinct genes but share a similar structure. α- and β-adducin include a protease-resistant N-terminal region and a protease-sensitive, hydrophilic C-terminal region. α- and γ-adducins are ubiquitously expressed. In contrast, β-adducin is expressed at high levels in brain and hematopoietic tissues. Adducin binds with high affinity to Ca(2+)/calmodulin and is a substrate for protein kinases A and C. Alternative splicing results in multiple variants encoding distinct isoforms; however, not all variants have been fully described.

Host Species: Rabbit; Mouse
Species Reactivity: Human

ELISA

Specifications

Antigen ADD1
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human ADD1.
Gene ADD1
Gene Alias ADDA/MGC3339/MGC44427
Gene Symbols ADD1
Host Species Mouse, Rabbit
Purification Method Affinity Purified
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 118
Includes Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-ADD1 (100 μg) 2. Detection antibody: mouse monoclonal anti-ADD1, IgG1 Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.