Learn More
Advanced ImmunoChemical, Inc. ANTI-ADIPONECTIN, HUMAN MAB
Supplier:  Advanced ImmunoChemical, Inc. 1ADP1 MG
Encompass_Preferred
Monoclonal Antibodies / Metabolic Syndrome/ Adiponectin. MAbs: Adn23, Adn27, Adn36, Adn63, Adn94, Adn214, Adn222, Adn279, Adn305. Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with human recombinant adiponectin from E. coli (MAbs Adn23) or with native human Monoclonal Antibodies. Metabolic Syndrome. Monoclonal Mouse Anti-Adiponectin (MAbs Adn27, Adn36, Adn63, Adn94, Adn214, Adn222, Adn279 and Adn305). Specificity: Human serum adiponectin.
Catalog No. NC3561639