Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADORA3, Mouse, Clone: 1A3, Abnova™

Catalog No. 89016057 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-057 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-057 Supplier Abnova Corporation Supplier No. H00000140M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ADORA3.

This gene encodes a protein that belongs to the family of adenosine receptors, which are G-protein-coupled receptors that are involved in a variety of intracellular signaling pathways and physiological functions. The receptor encoded by this gene mediates a sustained cardioprotective function during cardiac ischemia, it is involved in the inhibition of neutrophil degranulation in neutrophil-mediated tissue injury, it has been implicated in both neuroprotective and neurodegenerative effects, and it may also mediate both cell proliferation and cell death. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: VTTHRRIWLALGLCWLVSFLVGLTPMFGWNMKLTSEYHRNVTFLSCQFVSVMRMDYMVYFSFLTWIFIPLVVMCAIYLDIFYIIRNKLSLNLSNSKETGAFYGRE

Specifications

Antigen ADORA3
Applications ELISA
Classification Monoclonal
Clone 1A3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ADORA3.
Formulation PBS with no preservative; pH 7.4
Gene ADORA3
Gene Accession No. BC029831
Gene Alias A3AR/AD026/bA552M11.5
Gene Symbols ADORA3
Host Species Mouse
Immunogen ADORA3 (AAH29831, 121 a.a. ∼ 225 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.