Learn More
alpha-1d Adrenoceptor Polyclonal Antibody, Invitrogen™
Description
PA1-32690 detects alpha 1d Adrenoceptor from human samples.
Specifications
Specifications
| Antigen | alpha-1d Adrenoceptor |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | Adra1d |
| Gene Accession No. | P25100 |
| Gene Alias | [a]1d; ADA1D; Adra1; Adra-1; Adra1a; Adra1d; ADRA1R; Adrd1; adrenergic receptor delta1; adrenergic receptor, alpha 1d; adrenergic, alpha -1D-, receptor; adrenergic, alpha-1A-, receptor; adrenergic, alpha-1D-, receptor; adrenoceptor alpha 1D; alpha 1D-adrenoceptor; alpha 1D-adrenoreceptor; ALPHA1; alpha-1A adrenergic receptor; Alpha-1D adrenergic receptor; alpha-1D adrenoceptor; Alpha-1D adrenoreceptor; alpha-1D-adrenergic receptor; alpha1D-AR; Alpha1delta-AR; alpha-adrenergic receptor 1a; DAR; dJ779E11.2; Gpcr8; RA42; Spr8 |
| Gene Symbols | Adra1d |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.