Learn More
anti-alpha smooth muscle Actin, Clone: 1A4, Abcam™
This product is for research use only and not for diagnostic or therapeutic procedures. Customer is granted a limited, non-exclusive license for internal research and shall not attempt to reverse engineer.
Supplier: Abcam AB7817100UG

Description
Host Species: Mouse
Clone: 1A4
Isotype: IgG2a, κ
Cross Reactivity: Mouse, Rat, Human, Pig. Predicted to work with Rabbit, Cow, all Mammals, Baboon
Immunogen: The N-terminal decapeptide of alpha-smooth muscle actin.
Applications:
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blotting
Specifications
alpha smooth muscle Actin | |
1A4 | |
Unconjugated | |
The N-terminal decapeptide of alpha-smooth muscle actin. | |
Cardiovascular, Developmental Biology, Signal Transduction, Stem Cell Biology | |
Baboon, Bovine, Human, Mammalia, Mouse, Pig, Rabbit, Rat | |
IgG2a κ |
Monoclonal | |
0.2mg/mL | |
Mouse | |
500 μL | |
Primary | |
Supernatant |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.