Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyloid beta peptide (MOAB-2), pan Mouse anti-Human, Clone: 6C3, MilliporeSigma™

Catalog No. MABN254MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN254MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN254MI Supplier MilliporeSigma Supplier No. MABN254
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-amyloid beta peptide (MOAB-2), pan Antibody, clone 6C3 is an antibody against amyloid beta peptide (MOAB-2), pan, clone 6C3 for use in western blotting, immunohistochemistry (Paraffin), Immunofluorescence, immunoprecipitation, Dot Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen amyloid beta peptide (MOAB-2)
Applications Dot Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 6C3
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aλ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_000475
Gene Symbols APP, A4, AD1
Host Species Mouse
Immunogen Linear peptide corresponding to human pan amyloid beta peptide (MOAB-2).
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.