Learn More
ECM Bioscience Annexin A2 (C-terminal region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience AM2981
Encompass_Preferred
Clone (M298) was generated from a recombinant sequence that included amino acids in the C-terminal region of rat Annexin A2. Accession #Q07936. This antibody detects a 36 kDa* protein corresponding to the molecular mass of Annexin A2 on SDS-PAGE immunoblots of rat PC12 cells. The antibody has been shown to label Annexin A2 in immunocytochemical analysis of PC12 cells and immunohistochemical analysis of mouse diaphragm. Applications: WB, E, IP, ICC, IHC; Species Reactivity: Hu, Rt, Ms, Ck; Size = 100 μl; Buffer = Mouse monoclonal, protein A purified antibody is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at -20°C. Stable for 1 year. Other Names = Lipocortin, Calpactin 1, Annexin II
Catalog No. 50-206-8610
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.