Learn More
Description
Includes
Rabbit MaxPab™ affinity purified polyclonal anti-ANXA4 (100μg); Mouse purified polyclonal anti-ANXA4 (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Annexin IV (ANX4) belongs to the annexin family of calcium-dependent phospholipid binding proteins. Although their functions are still not clearly defined, several members of the annexin family have been implicated in membrane-related events along exocytotic and endocytotic pathways. ANX4 has 45 to 59% identity with other members of its family and shares a similar size and exon-intron organization. Isolated from human placenta, ANX4 encodes a protein that has possible interactions with ATP, and has in vitro anticoagulant activity and also inhibits phospholipase A2 activity. ANX4 is almost exclusively expressed in epithelial cells.
ELISA
Specifications
Specifications
| Antigen | ANXA4 |
| Applications | ELISA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Description | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human ANXA4. |
| Gene | ANXA4 |
| Gene Alias | ANX4/DKFZp686H02120/MGC75105/PIG28/ZAP36 |
| Gene Symbols | ANXA4 |
| Host Species | Mouse, Rabbit |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.