Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apolipoprotein E Goat anti-Human, Primate, Polyclonal, MilliporeSigma™

Catalog No. AB947MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB947MI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB947MI Supplier MilliporeSigma Supplier No. AB947
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Detect Apolipoprotein E using this Anti-Apolipoprotein E Antibody validated for use in immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apolipoprotein E
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Delipidated goat antiSerum, 0.2 μm filtered. Liquid in Tris-saline, pH 7.4 with 0.1% sodium azide as preservative.
Gene Symbols APOE; Apo-E; apoprotein; MGC1571; AD2
Host Species Goat
Immunogen Recombinant human apolipoprotein E.
Purification Method Unpurified
Quantity 1 mL
Primary or Secondary Primary
Gene ID (Entrez) NM_000041.2
Target Species Human, Primate
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.