Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APP A4 Mouse anti-Canine, Human, Mouse, Monkey, Pig, Rat, Fish, Clone: 22C11, MilliporeSigma™

Catalog No. mab348mi Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB348MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB348MI Supplier MilliporeSigma Supplier No. MAB348
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-APP A4 Antibody, a.a. 66-81 of APP {N-terminus}, clone 22C11 is an antibody against Alzheimer Precursor Protein A4 for use in immunofluoresence, immunohistochemistry, immunohistochemistry (paraffin) and western blotting with more than 85 product citations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APP A4
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 22C11
Conjugate Unconjugated
Formulation Protein A Purified mouse immunoglobulin in 20mM sodium phosphate, 250mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative.
Gene Accession No. NM_000484.2; NM_201413.1; NM_201414.1
Gene Symbols APP, PN-II, AD1, ABETA, CTFgamma, APPI, ABPP, A4, CVAP, PN2, PreA4, AAA
Host Species Mouse
Immunogen Purified recombinant Alzheimer precursor A4 (pre A4695) fusion protein.
Purification Method Protein A purfied
Quantity 50 μg
Primary or Secondary Primary
Target Species Canine, Fish, Human, Monkey, Mouse, Pig, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.