Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arginine Vasopressin Receptor V2 Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB1797PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1797PMI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1797PMI Supplier MilliporeSigma Supplier No. AB1797P
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Arginine Vasopressin Receptor V2 Antibody is an antibody against Arginine Vasopressin Receptor V2 for use in ELISA, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Arginine Vasopressin Receptor V2
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in PBS containing 0.1% BSA.
Gene Symbols AVPR2; DI1; V2R; DIR; MGC138386; MGC126533; DIR3; ADHR; NDI
Host Species Rabbit
Immunogen 21 amino acid peptide from the 3rd cytoplasmic domain of rat AVP-V2 (Lolait et al. 1992; Fisov et al. 1994).
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_000054.2
Target Species Rat
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.