Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rho guanine nucleotide exchange factor (GEF) 1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004423 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-423 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-423 Supplier Abnova Corporation Supplier No. H00009138A01
Only null left
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant ARHGEF1.

Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form complex with G proteins and stimulate Rho-dependent signals. Multiple alternatively spliced transcript variants have been found for this gene, but the full-length nature of some variants has not been defined. [provided by RefSeq

Sequence: CRPGPEGQLAATALRKVLSLKQLLFPAEEDNGAGPPRDGDGVPGGGPLSPARTQEIQENLLSLEETMKQLEELEEEFCRLRPLLSQLGGNSVPQPGCT

Specifications

Antigen Rho guanine nucleotide exchange factor (GEF) 1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ARHGEF1.
Formulation 50% glycerol
Gene ARHGEF1
Gene Accession No. BC034013
Gene Alias GEF1/LBCL2/LSC/P115-RHOGEF/SUB1.5
Gene Symbols ARHGEF1
Host Species Mouse
Immunogen ARHGEF1 (AAH34013, 830 a.a. ∼ 927 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cell Cycle
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 9138
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.