Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rho guanine nucleotide exchange factor (GEF) 7, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004410 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-410 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-410 Supplier Abnova Corporation Supplier No. H00008874A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant ARHGEF7.

Rho GTPases play a fundamental role in numerous cellular processes triggered by extracellular stimuli that work through G protein coupled receptors. The encoded protein belongs to a family of cytoplasmic proteins that activate the Ras-like family of Rho proteins by exchanging bound GDP for GTP. It forms a complex with the small GTP binding protein Rac1 and recruits Rac1 to membrane ruffles and to focal adhesions. This protein can induce membrane ruffling. Multiple alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq

Sequence: SSLVTLNKVTADIGLGSDSVCARPSSHRIKSFDSLGSQSLHTRTSKLFQGQYRSLDMTDNSNNQLVVRAKFNFQQTNEDELSFSKGDVI

Specifications

Antigen Rho guanine nucleotide exchange factor (GEF) 7
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ARHGEF7.
Formulation 50% glycerol
Gene ARHGEF7
Gene Accession No. NM_145735
Gene Alias BETA-PIX/COOL1/DKFZp686C12170/DKFZp761K1021/KIAA0142/KIAA0412/Nbla10314/P50/P50BP/P85/P85COOL1/P85SPR/PAK3/PIXB
Gene Symbols ARHGEF7
Host Species Mouse
Immunogen ARHGEF7 (NP_663788, 102 a.a. ∼ 190 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 8874
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.