Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATM (pS1981) Mouse, Unlabeled, Clone: K88-534, BD

Catalog No. BDB560007 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560007 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560007 Supplier BD Biosciences Supplier No. 560007
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ATM is the gene that is mutated in ataxia telangiectasia, an autosomal recessive hereditary disease that is characterized by delayed motor and sexual development, weakened immunity, multiple skin changes, increased sensitivity to ionizing radiation, and increased risk of developing certain cancers. The protein encoded by ATM is a phophoinositide-3-kinase-related protein kinase that is found in the nucleus and is responsible for early responses to the changes in chromatin structure caused by DNA double-strand breaks. Inactive wild type ATM protein forms homodimers or higher order multimers in which the C-terminal kinase domains are inhibited. It has been proposed that multimeric ATM is recruited to DNA double-strand breaks through interactions between its N-terminal HEAT domains and specific chromatin protein complexes. The inhibition of the kinase domains is lifted, resulting in auto-phosphorylation of the ATM at a single serine site and disruption of the multimers. The phosphorylated state of ATM is a rapid and highly sensitive indicator that cells have been exposed to agents that cause DNA double-strand breaks. The kinase domains of the resulting ATM monomers are accessible to a wide variety of substrates that are involved in DNA repair, cell-cycle regulation, and apoptosis. Thus ATM is a central regulator of cellular responses to ionizing radiation, and cells that lack ATM undergo radioresistant DNA synthesis and are resistant to γradiation-induced apoptosis .

The K88-534 monoclonal antibody recognizes human ATM phosphorylated at serine 1981 (S1981) and mouse ATM phosphorylated at its orthologous site, S1987.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ATM (pS1981)
Applications In vitro Assay
Classification Monoclonal
Clone K88-534
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Mouse ATM Peptide
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.