Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATPase, H+ transporting, lysosomal 42kDa, V1 subunit C2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018695 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-695 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-695 Supplier Abnova Corporation Supplier No. H00245973D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human ATP6V1C2 protein.

This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A, three B, and two G subunits, as well as a C, D, E, F, and H subunit. The V1 domain contains the ATP catalytic site. This gene encodes alternate transcriptional splice variants, encoding different V1 domain C subunit isoforms. [provided by RefSeq

Sequence: MSEFWLISAPGDKENLQALERMNTVTSKSNLSYNTKFAIPDFKVGTLDSLVGLSDELGKLDTFAESLIRRMAQSVVEVMEDSKGKVQEHLLANGVDLTSFVTHFEWDMAKYPVKQPLVSVVDTIAKQLAQIEMDLKSRTAAYDTLKTNLENLEKKSMGNLFTRTLSDIVSKEDFVLDSEYLVTLLVIVPKPNYSQWQKTYESLSDMVVPRSTKLITEDKEGGLFTVTLFRKVIEDFKTKAKENKFTVREFYYDEKEIEREREEMARLLSDKKQQYGPLLRWLKVNFSEAFIAWIHIKALRVFVESVLRYGLPVNFQAVLLQPHKKSSTKRLREVLNSVFRHLDEVAATSILDASVEIPGLQLNNQDYFPYVYFHIDLSLLD

Specifications

Antigen ATPase, H+ transporting, lysosomal 42kDa, V1 subunit C2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human ATP6V1C2 protein.
Formulation PBS with no preservative; pH 7.4
Gene ATP6V1C2
Gene Accession No. BC012142.1
Gene Alias ATP6C2/VMA5
Gene Symbols ATP6V1C2
Host Species Rabbit
Immunogen ATP6V1C2 (AAH12142.1, 1 a.a. ∼ 381 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245973
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.