Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Src Mouse anti-Avian, Chicken, Clone: EC10, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05185I100UG
Description
Src Monoclonal antibody specifically detects Src in Avian, Chicken samples. It is validated for Immunofluorescence, Immunoprecipitation, Western Blotting.
Specifications
| avian Src | |
| Monoclonal | |
| 1.0 mg/mL | |
| Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
| pp60src from SR-1T cells. | |
| 100 μg | |
| Primary | |
| Stable for 1 year at +2°C to +8°C from date of receipt. | |
| IgG2b κ |
| Immunofluorescence, Immunoprecipitation, Western Blot | |
| EC10 | |
| Unconjugated | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Avian, Chicken | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction