Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AXIN1 up-regulated 1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004708 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-708 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-708 Supplier Abnova Corporation Supplier No. H00064651A01
Only null left
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant AXUD1.

This gene encodes a protein that localizes to the nucleus and expression of this gene is induced in response to elevated levels of axin. The Wnt signalling pathway, which is negatively regulated by axin, is important in axis formation in early development and impaired regulation of this signalling pathway is often involved in tumors. A decreased level of expression of this gene in tumors compared to the level of expression in their corresponding normal tissues suggests that this gene product has a tumor suppressor function. [provided by RefSeq

Sequence: MTGLLKRKFDQLDEDNSSVSSSSSSSGCQSRSCSPSSSVSRAWDSEEEGPWDQMPLPDRDFCGPRSFTPLSILKRARRERPGRVAFDGITVFYFPRCQGFTSVPSRGGCTLGMALRHSACRRFSLAEFAQEQARARHEKLRQRLKEEKLEMLQWKLSAAGVPQAEAGLPPVVDAIDDASVEEDLAVAVAGGRLEEVSFLQPYPARRRRALLRASGVRRIDREEKRELQALRQSREDCGCHCDRICDPETCSCSLAGIKCQMDHTAFPCGCCREGCENPMGRVEFNQARVQTHFIHTLTRLQLEQEAESFRELEAPAQGSPPSPGEEALVPTFPLAKPPMNNELGDNSCSSDMTDSSTASSSASGTSEAPDCPTHPGLPGPGFQPGVDDDSLARILSFSDSDFGGEEEEEEEGSVGNLDNLSCFHPADIFGTSDPGGLASWTHSYSGCSFTSGILDENANLDASCFLNGGLEGSREGSLPGTSVPPSMDAGRSSSVDLSLSSCDSFELLQALPDYSLGPHYTSQKVSDSLDNIEAPHFPLPGLSPPGDASSCFLESLMGFSEPAAEALDPFIDSQFEDTVPASLMEPVPV

Specifications

Antigen AXIN1 up-regulated 1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant AXUD1.
Formulation 50% glycerol
Gene AXUD1
Gene Accession No. BC038949
Gene Alias CSRNP1/DKFZp566F164/FAM130B/TAIP-3/URAX1
Gene Symbols AXUD1
Host Species Mouse
Immunogen AXUD1 (AAH38949, 1 a.a. ∼ 589 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Apoptosis
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 64651
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.