Learn More
ECM Bioscience B7-H3/CD276 (Extracellular region) Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience BM0451
Encompass_Preferred
Clone (M045) was generated from a recombinant protein that included the extracellular region of human B7-H3 protein. Accession #Q5ZPR3. Clone M045 mouse monoclonal antibody detects a 100-120 kDa* protein on SDS-PAGE "Native" or denatured immunoblots of human LNCaP, MCF-7, NCI-H28, A431, and MDA-MB-231 carcinomas. The antibody detects B7-H3 in membranes and cytoplasm in MCF7 cell after immunocytochemical labeling. The antibody works for western blot, immunoprecipitation, immunocytochemistry, and ELISA capture. Applications: WB, EC, IP, ICC; Species Reactivity: Hu; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein G chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = CD276, B7-H3, B7H3, B7RP-2, B7 homolog 3
Catalog No. 50-206-8618