Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

bromodomain adjacent to zinc finger domain, 1B, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004417 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-417 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-417 Supplier Abnova Corporation Supplier No. H00009031A01
Only null left
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant BAZ1B.

This gene encodes a member of the bromodomain protein family. The bromodomain is a structural motif characteristic of proteins involved in chromatin-dependent regulation of transcription. This gene is deleted in Williams-Beuren syndrome, a developmental disorder caused by deletion of multiple genes at 7q11.23. [provided by RefSeq

Sequence: MDFQTVQNKCSCGSYRSVQEFLTDMKQVFTNAEVYNCRGSHVLSCMVKTEQCLVALLHKHLPGHPYVRRKRKKFPDRLAEDEGDSEPEAVGQSRGRRQKK

Specifications

Antigen bromodomain adjacent to zinc finger domain, 1B
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant BAZ1B.
Formulation 50% glycerol
Gene BAZ1B
Gene Accession No. NM_023005
Gene Alias WBSCR10/WBSCR9/WSTF
Gene Symbols BAZ1B
Host Species Mouse
Immunogen BAZ1B (NP_075381, 1384 a.a. ∼ 1483 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Transcription Regulation
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 9031
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.