Learn More
Description
Expression of Bcl-2α oncoprotein inhibits the programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of Bcl-2α protein, whereas the normal or hyperplastic germinal centers are negative.
Host Species: Mouse
Clone: 10C4
Isotype: IgG1
Cross Reactivity: Mouse and Rat. Does not react with human. Others-not tested.
Epitope: aa 61-76
Immunogen: A Synthetic peptide, aa 61-76 (Cys-VHREMAARTSPLRPLV), of Mouse bcl-2 protein.
Molecular Weight: 25-26kDa
Positive Control: MAD109 cells.
Cellular Localization: Cytoplasm
Ab-4 is particularly designed for research on mouse and rat bcl-2alpha protein. It does not react with human bcl-2alpha protein.
Immunofluorescence, Immunoprecipitation, Western Blotting
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
| Antigen | BCL-2α Ab-4 |
| Classification | Monoclonal |
| Concentration | 200μg/mL |
| Host Species | Mouse |
| Immunogen | A Synthetic peptide, aa 61-76 (Cys-VHREMAARTSPLRPLV), of Mouse bcl-2 protein. |
| Quantity | 1 mL |
| Primary or Secondary | Primary |
| Target Species | Mouse, Rat |
| Isotype | IgG1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.