Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-2 w/control Mouse anti-Human, FITC, Clone: BCL-2/100, BD

Catalog No. BDB556357 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556357 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556357 Supplier BD Biosciences Supplier No. 556357
Only null left
Add to Cart
Add to Cart

Includes

FITC Mouse Anti-Human Bcl-2, FITC Mouse IgG1, κ Isotype Control

Mouse Monoclonal Antibody

Programmed cell death (apoptosis) is a normal physiologic process which occurs during embryonic development as well as in maintenance of tissue homeostasis. The apoptotic program is characterized by certain morphological features. These include changes in the plasma membrane such as loss of membrane asymmetry and attachment, a condensation of the cytoplasm and nucleus, and internucleosomal cleavage of DNA. In the final stages, the dying cells become fragmented into “apoptotic bodies” which are rapidly eliminated by phagocytic cells without eliciting significant inflammatory damage to surrounding cells. Members of the Bcl-2 family play a major role in regulating the response of cells to apoptotic signals. Bcl-2 is considered to be novel among proto-oncogenes because it blocks apoptosis in many cell types. Bcl-2 is thought to provide selective survival advantage for cells by blocking apoptosis and thus may contribute to tumorigenesis. Bcl-2 is a 26 kDa intracellular, integral membrane protein found primarily in the nuclear envelope, endoplasmic reticulum and outer mitochondrial membrane.

Clone Bcl-2/100 reacts with human Bcl-2. It does not cross-react with mouse Bcl-2. A synthetic peptide corresponding to amino acids 41-54 (GAAPAPGIFSSQPG) of human Bcl-2 was used as immunogen. This peptide sequence is not conserved between human and mouse. Clone Bcl-2/100 was initially characterized by western blotting and immunohistochemistry of frozen and paraffin-embedded tissue sections. Clone MOPC-21 is a mouse IgG1 isotype control. The MOPC-21 antibody has unknown specificity. The Bcl-2/100 and MOPC-21 FITC conjugates are matched in F/P ratios. The optimum F/P ratio was experimentally determined by flow cytometric analysis.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-2 w/control
Applications Flow Cytometry
Classification Monoclonal
Clone BCL-2/100
Conjugate FITC
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Bcl-2 synthetic peptide aa. 41-54
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Includes FITC Mouse Anti-Human Bcl-2 (100 Tests), FITC Mouse IgG1, κ Isotype Control (100 Tests)
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.