Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-BCR, PIK3CG Protein Protein Interaction Antibody Pair, Abnova™

Catalog No. 89003532 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-003-532 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-003-532 Supplier Abnova Corporation Supplier No. DI0613
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

BCR rabbit purified polyclonal antibody (20μg); PIK3CG mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols.

Host Species: Rabbit, Mouse
Cross Reactivity: Human

In situ PLA

Specifications

Antigen BCR
Applications In situ PLA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Gene BCR
Gene Alias ALL/BCR-ABL1/BCR1/CML/D22S11/D22S662/FLJ16453/PHL
Gene Symbols BCR
Host Species Mouse, Rabbit
Quantity 1 Set
Primary or Secondary Primary
Gene ID (Entrez) 613/5294
Includes Antibody pair set content: 1. BCR rabbit purified polyclonal antibody (20 ug) 2. PIK3CG mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Product Type BCR & PIK3CG Protein Protein Interaction Antibody Pair
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.