Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Betacellulin/BTC Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2611
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2611 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB2611 Supplier R&D Systems Supplier No. MAB2611
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Betacellulin/BTC Monoclonal specifically detects Betacellulin/BTC in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Betacellulin/BTC
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 34301
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P35070
Gene Alias betacellulin, BTC, probetacellulin
Gene Symbols BTC
Host Species Mouse
Immunogen E. coli-derived recombinant human Betacellulin/BTC Asp32-Tyr111 Accession # P35070
Purification Method Protein A or G purified from ascites
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 685
Test Specificity Detects human Betacellulin/BTC in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) BTC, recombinant human (rh) Amphiregulin, rhEGF, rhHB-EGF, rhHeregulin-alpha , rhHeregulin-beta , rhTGF-alpha , or rmEpiregulin is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.