Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat Anti-Blimp-1 Rat, PE, Clone: 6D3, BD

Catalog No. BDB564702 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564702 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564702 Supplier BD Biosciences Supplier No. 564702
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The 6D3 monoclonal antibody specifically binds to mouse B lymphocyte-induced maturation protein 1 (Blimp-1) and crossreacts with human BLIMP-1. Blimp-1 is a 98 kDa zinc finger-containing protein that is encoded by the Prdm1 gene. Blimp-1 was first discovered as a transcriptional repressor of the IFN- β promoter. Blimp-1 is critical for primordial germ cells specification in the early embryo and induction of terminal cell differentiation in multiple somatic cell types. It is largely known as a master regulator of antibody-secreting plasma cells due to its function in the repression of PAX5 , CIITA and cMYC genes. Blimp-1 also controls T cell homeostasis and effector differentiation through different mechanisms, including repression of IL-17 producing cells. Blimp-1 also plays roles in the maturation and activity of dendritic cells and natural killer cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat Anti-Blimp-1
Applications Flow Cytometry
Classification Monoclonal
Clone 6D3
Concentration 0.2mg/mL
Conjugate PE
Description BLIMP-1; PRDM1; PR domain containing 1, with ZNF domain; PRDI-BF1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse Blimp-1 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.