Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMP5, Mouse, Clone: 1C1, Abnova™

Catalog No. 89016328 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-328 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-328 Supplier Abnova Corporation Supplier No. H00000653M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant BMP5.

This gene encodes a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. These proteins are synthesized as prepropeptides, cleaved, and then processed into dimeric proteins. This protein may act as an important signaling molecule within the trabecular meshwork and optic nerve head, and may play a potential role in glaucoma pathogenesis. This gene is differentially regulated during the formation of various tumors. [provided by RefSeq

Sequence: VGDYNTSEQKQACKKHELYVSFRDLGWQDWIIAPEGYAAFYCDGECSFPLNAHMNATNHAIVQTLVHLMFPDHVPKPCCAPTKLNAISVLYFDDSSNVIL

Specifications

Antigen BMP5
Applications ELISA
Classification Monoclonal
Clone 1C1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant BMP5.
Formulation PBS with no preservative; pH 7.4
Gene BMP5
Gene Accession No. NM_021073.1
Gene Alias MGC34244
Gene Symbols BMP5
Host Species Mouse
Immunogen BMP5 (NP_066551.1, 341 a.a. ∼ 440 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 653
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.