Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BrdU with DNase Mouse anti-Human, FITC, Clone: B44, BD

Catalog No. BDB340649 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB340649 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB340649 Supplier BD Biosciences Supplier No. 340649
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The Anti-BrdU antibody, clone B44, is derived from hybridization of Sp2/0-Ag14 mouse myeloma cells with spleen cells from BALB/c mice immunized with iodouridine-conjugated ovalbumin.

Bromodeoxyuridine (BrdU) is a uridine derivative that can be incorporated specifically into DNA in place of thymidine. Anti-BrdU identifies BrdU (but not thymidine) in single-stranded DNA, free BrdU, or BrdU coupled to a protein carrier. The antibody also reacts with iodouridine. DNase-I cleaves each strand of DNA at random and permits the binding of Anti-BrdU antibody to its antigen (BrdU).

TRUSTED_SUSTAINABILITY

Specifications

Antigen BrdU with DNase
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone B44
Concentration 10μg/mL
Conjugate FITC
Formulation Balanced salt solution with DNase, gelatin, and 0.1% sodium azide.
Host Species Mouse
Immunogen Iodouridine
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.