Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BZRP, Mouse, Clone: 3D8-B2, Abnova™

Catalog No. 89016375 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-375 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-375 Supplier Abnova Corporation Supplier No. H00000706M01
Only null left
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant BZRP.

Present mainly in the mitochondrial compartment of peripheral tissues, the protein encoded by this gene interacts with some benzodiazepines and has different affinities than its endogenous counterpart. The protein is a key factor in the flow of cholesterol into mitochondria to permit the initiation of steroid hormone synthesis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq

Sequence: MAPPWVPAMGFTLAPSLGCFVGSRFVHGEGLRWYAGLQKPSWHPPHWVLGPVWGTLYSAMGYGSYLVWKELGGFTEKAVVPLGLYTGQLALNWAWPPIFFGARQMGWALVDLLLVSGAAAATTVAWYQVSPLAARLLYPYLAWLAFATTLNYCVWRDNHGWHGGRRLPE

Specifications

Antigen BZRP
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3D8-B2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant BZRP.
Formulation PBS with no preservative; pH 7.4
Gene TSPO
Gene Accession No. BC001110
Gene Alias BZRP/DBI/IBP/MBR/PBR/PKBS/PTBR/mDRC/pk18
Gene Symbols TSPO
Host Species Mouse
Immunogen BZRP (AAH01110.1, 1 a.a. ∼ 169 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 706
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.