Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Cbl (pY700) Mouse anti-Human, Unlabeled, Clone: 47/C-CBL, BD

Catalog No. p-3879071 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612304 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612304 Supplier BD Biosciences Supplier No. 612304

Mouse Monoclonal Antibody

Cbl (Casitas B-lineage lymphoma) was identified in the genome of a transforming retrovirus from a mouse pre-B lymphoma. The cellular gene product c-Cbl is one of numerous Cbl-related proteins found in vertebrate and invertebrate organisms. It is an 120kDa adapter protein that contains multiple functional domains, including a RING finger motif, a tyrosine kinase-binding (TKB) domain, and a proline-rich region. The TKB domain directly interacts with specific auto-phosphorylation sites in activated protein-tyrosine kinases (PTK). Through the RING finger motif, c-Cbl recruits and activates an E2 ubiquitin-conjugating enzyme, thus targeting the activated PTK for protein degradation. The proline-rich region contains SH3 domain-binding and 14-3-3 protein-binding motifs. c-Cbl is also phosphorylated at tyrosines 700 (Y700), 731, and 774 by Syk- and Src-family kinases after the stimulation of some integrins and a wide variety of receptors for antigens, immunoglobulins, growth factors, cytokines, and hormones. In turn, the phosphorylated Y700 site interacts with the SH2 domains of CRK and Vav1. The c-Cbl adapter protein is expressed in the cytoplasm in all tissues, with especially high levels of expression in hematopoietic cells. Through its many functional sites, c-Cbl plays key roles in the positive and negative regulation of vital cell functions, including T Cell Receptor-mediated cellular immune responses.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen c-Cbl (pY700)
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 47/C-CBL
Concentration 250μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human c-Cbl (pY700) Peptide
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.