Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

c-Raf Mouse, Unlabeled, Clone: 53, BD

Catalog No. p-3855690 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610151 50 μg
BDB610152 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610151 Supplier BD Biosciences Supplier No. 610151
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Raf (c-Raf-1) is a cytoplasmic serine/threonine protein kinase and is a member of a family of proteins which are highly conserved from Drosophila to mammals. Raf has a critical role in the response to many growth factors including: EGF, PDGF, insulin, IL-2, IL-3, CSF-1, and GM-CSF. Raf can directly interact with Ras-GTP and subsequently become activated. Thus Raf plays a prominent role in the Ras signaling pathway by transferring a signal from Ras which has been activated by growth factor receptor-stimulated tyrosine kinase activity. This leads to the stimulation and activation of a number of other cytoplasmic serine/threonine kinases. Raf regulates the MAP kinase pathway by phosphorylating and activating MEK, which then phosphorylates and activates MAP kinase (ERK). ERK then phosphorylates and activates Rsk. Raf activity can also be regulated independently of Ras. Cyclic AMP (cAMP) activation of protein kinase A (PKA) can inhibit growth factor stimulation of Raf.

TRUSTED_SUSTAINABILITY

Specifications

Antigen c-Raf
Applications Western Blot
Classification Monoclonal
Clone 53
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.